Bis(tert-butylcyclopentadienyl)hafnium dichloride ≥98%

Supplier: Thermo Scientific
Warning

H27638.03 H27638.06
ALFAH27638.03EA 0 SGD
ALFAH27638.03 ALFAH27638.06
Bis(tert-butylcyclopentadienyl)hafnium dichloride ≥98%
Bis(tert-butylcyclopentadienyl)hafnium dichloride

Purity: 98+% CAS No.: 33010-55-8 Molecular Formula: C18H26Cl2Hf Formula Weight: 491.80 Melting Point: 195° MDL No.: MFCD01073803

Formula: C₁₈H₂₆Cl₂Hf
MW: 491,80 g/mol
Melting Pt: 195 °C
Storage Temperature: Ambient
MDL Number: MFCD01073803
CAS Number: 33010-55-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR