Methyl-4-(chloromethyl)benzoate ≥97%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Methyl 4-(chloromethyl)benzoate

B22476.04 B22476.09
ALFAB22476.04EA 0 SGD
ALFAB22476.04 ALFAB22476.09
Methyl-4-(chloromethyl)benzoate ≥97%
Methyl 4-(chloromethyl)benzoate

Purity: 97% CAS No.: 34040-64-7 Molecular Formula: C9H9ClO2 Formula Weight: 184.62 Melting Point: 39-40° Boiling Point: 160-165°/30mm UN No.: 3261 MDL No.: MFCD00511224

Formula: C₉H₉ClO₂
MW: 184.62 g/mol
Boiling Pt: 160…165 °C (30 mmHg)
Melting Pt: 39…40 °C
Storage Temperature: Ambient
MDL Number: MFCD00511224
CAS Number: 34040-64-7
EINECS: 251-806-6
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR