4'-Methoxybiphenyl-4-ylboronic acid pinacol ester 95%
Supplier: Thermo Scientific
Synonyms:
2-[4-(4-Methoxyphenyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-(4'-Methoxy-4-biphenylyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
H51907.03
H51907.06
ALFAH51907.03EA
122
SGD
ALFAH51907.03
ALFAH51907.06
4'-Methoxybiphenyl-4-ylboronic acid pinacol ester 95%
4'-Methoxybiphenyl-4-ylboronic acid pinacol ester
Purity: 95%
Molecular Formula: C19H23BO3
Formula Weight: 310.20
MDL No.: MFCD16294530
Formula:
C₁₉H₂₃BO₃ MW: 310,20 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16294530 CAS Number: 446311-34-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...