4-(4-Hydroxyphenyl)benzonitrile ≥99%

Supplier: Thermo Scientific
Danger

Synonyms: 4'-Hydroxy-4-biphenylcarbonitrile, 4'-Hydroxybiphenyl-4-carbonitrile, 4'-Hydroxy-[1,1'-biphenyl]-4-carbonitrile

B21864.03 B21864.06
ALFAB21864.03EA 0 SGD
ALFAB21864.03 ALFAB21864.06
4-(4-Hydroxyphenyl)benzonitrile ≥99%
4-(4-Hydroxyphenyl)benzonitrile

Purity: 99% CAS No.: 19812-93-2 Molecular Formula: C13H9NO Formula Weight: 195.22 Melting Point: 196-199° UN No.: 3439 MDL No.: MFCD00059625

Formula: C₁₃H₉NO
MW: 195,22 g/mol
Melting Pt: 196…199 °C
Storage Temperature: Ambient
MDL Number: MFCD00059625
CAS Number: 19812-93-2
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR