2-Methoxynicotinonitrile ≥97%

Supplier: Thermo Scientific
Danger

Synonyms: 3-Cyano-2-methoxypyridine, 2-Methoxynicotinonitrile

H52373.MD H52373.03
ALFAH52373.MDEA 0 SGD
ALFAH52373.MD ALFAH52373.03
2-Methoxynicotinonitrile ≥97%
2-Methoxynicotinonitrile 2-(4-Formylphenyl)isonicotinonitrile

Purity: 97+% CAS No.: 7254-34-4 Molecular Formula: C7H6N2O Formula Weight: 134.14 MDL No.: MFCD07363804

Formula: C₇H₆N₂O
MW: 134,14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07363804
CAS Number: 7254-34-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR