4-Amino-3,5-dimethyl-1,2,4-triazole 98%

Supplier: Thermo Scientific
Warning

B20417.03 B20417.06
ALFAB20417.03EA 0 SGD
ALFAB20417.03 ALFAB20417.06
4-Amino-3,5-dimethyl-1,2,4-triazole 98%
4-Amino-3,5-dimethyl-1,2,4-triazole

Purity: 98% CAS No.: 3530-15-2 Molecular Formula: C4H8N4 Formula Weight: 112.14 Melting Point: 196-199° Beilstein Reference: 115482

Formula: C₄H₈N₄
MW: 112,14 g/mol
Melting Pt: 196…199 °C
Storage Temperature: Ambient
MDL Number: MFCD00051647
CAS Number: 3530-15-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR