4-(Furfurylcarbamoyl)phenylboronic acid pinacol ester 97%

Supplier: Thermo Scientific

Synonyms: N-(Furan-2-ylmethyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide, 4-(Furfurylaminocarbonyl)benzeneboronic acid pinacol ester

H29236.03 H29236.06
ALFAH29236.03EA 0 SGD
ALFAH29236.03 ALFAH29236.06
4-(Furfurylcarbamoyl)phenylboronic acid pinacol ester 97%
4-(Furfurylcarbamoyl)phenylboronic acid pinacol ester

Purity: 97% CAS No.: 1073353-59-9 Molecular Formula: C18H22BNO4 Formula Weight: 327.18 Melting Point: 113-114° MDL No.: MFCD09266168

Formula: C₁₈H₂₂BNO₄
MW: 327,18 g/mol
Melting Pt: 113…114 °C
Storage Temperature: Ambient
MDL Number: MFCD09266168
CAS Number: 1073353-59-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR