4-(4-Morpholinylcarbonyl)phenylboronic acid pinacol ester ≥97%

Supplier: Thermo Scientific

H28484.06 H28484.03
ALFAH28484.06EA 0 SGD
ALFAH28484.06 ALFAH28484.03
4-(4-Morpholinylcarbonyl)phenylboronic acid pinacol ester ≥97%
4-(4-Morpholinylcarbonyl)phenylboronic acid pinacol ester

Purity: 97% CAS No.: 656239-38-2 Molecular Formula: C17H24BNO4 Formula Weight: 317.19 Melting Point: 187-193° MDL No.: MFCD05863913

Formula: C₁₇H₂₄BNO₄
MW: 317,19 g/mol
Melting Pt: 187…193 °C
Storage Temperature: Ambient
MDL Number: MFCD05863913
CAS Number: 656239-38-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR