5-(1-Piperidinylmethyl)thiophene-2-boronic acid pinacol ester
Supplier: Thermo Scientific
Synonyms:
1-{[5-(Tetramethyl-1,3,2-dioxaborolan-2-yl)thiophen-2-yl]methyl}piperidine
H51117.03
H51117.06
ALFAH51117.03EA
0
SGD
ALFAH51117.03
ALFAH51117.06
5-(1-Piperidinylmethyl)thiophene-2-boronic acid pinacol ester
5-(1-Piperidinylmethyl)thiophene-2-boronic acid pinacol ester
CAS No.: 1218790-44-3
Molecular Formula: C16H26BNO2S
Formula Weight: 307.26
MDL No.: MFCD11113036
Formula:
C₁₆H₂₆BNO₂S MW: 307,26 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11113036 CAS Number: 1218790-44-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...