2,2'-Thenoin ≥97%
Supplier: Thermo Scientific
B24671.03
B24671.06
ALFAB24671.03EA
0
SGD
ALFAB24671.03
ALFAB24671.06
2,2'-Thenoin ≥97%
2,2'-Thenoin
Purity: 97%
CAS No.: 27761-02-0
Molecular Formula: C10H8O2S2
Formula Weight: 224.29
Melting Point: 104-107°
MDL No.: MFCD00022505
Formula:
C₁₀H₈O₂S₂ MW: 224,29 g/mol Melting Pt: 104…107 °C Storage Temperature: Ambient |
MDL Number:
MFCD00022505 CAS Number: 27761-02-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...