4-(3-Piperidinyloxy)benzonitrile ≥97%
Supplier: Thermo Scientific
|
Warning
|
H50995.03
H50995.MD
ALFAH50995.03EA
495
SGD
ALFAH50995.03
ALFAH50995.MD
4-(3-Piperidinyloxy)benzonitrile ≥97%
4-(3-Piperidinyloxy)benzonitrile
Purity: 97%
CAS No.: 902837-34-7
Molecular Formula: C12H14N2O
Formula Weight: 202.26
MDL No.: MFCD08061105
Formula:
C₁₂H₁₄N₂O MW: 202,26 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08061105 CAS Number: 902837-34-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...