(R)-(-)-3-Aminopyrrolidine dihydrochloride 97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
(R)-Pyrrolidin-3-amine dihydrochloride
L11173.03
L11173.MD
ALFAL11173.03EA
0
SGD
ALFAL11173.03
ALFAL11173.MD
(R)-(-)-3-Aminopyrrolidine dihydrochloride 97%
(R)-(-)-3-Aminopyrrolidine dihydrochloride
Purity: 98%
CAS No.: 116183-81-4
Molecular Formula: C4H10N2.2HCl
Formula Weight: 159.06
Melting Point: ca 283° dec.
Beilstein Reference: 5852700
Formula:
C₄H₁₂Cl₂N₂ MW: 159,06 g/mol Melting Pt: ∼ 283 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00070604 CAS Number: 116183-81-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...