1-Boc-4-iodopiperidine 95%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
1-BOC-4-iodo-piperidine, tert-Butyl 4-iodopiperidine-1-carboxylate
H34090.03
H34090.06
ALFAH34090.03EA
0
SGD
ALFAH34090.03
ALFAH34090.06
1-Boc-4-iodopiperidine 95%
1-Boc-4-iodopiperidine
Purity: 95%
CAS No.: 301673-14-3
Molecular Formula: C10H18INO2
Formula Weight: 311.16
Storage Temperature: 2°C to 8°C
Formula:
C₁₀H₁₈INO₂ MW: 311,16 g/mol |
MDL Number:
MFCD04115041 CAS Number: 301673-14-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...