3-Bromo-4-hydroxy-5-methoxycinnamic acid 97

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 3-(3-Bromo-4-hydroxy-5-methoxyphenyl)acrylic acid(predominantly trans), 3-(3-Bromo-4-hydroxy-5-methoxyphenyl)acrylic acid

B20014.03 B20014.06
ALFAB20014.03EA 0 SGD
ALFAB20014.03 ALFAB20014.06
3-Bromo-4-hydroxy-5-methoxycinnamic acid 97
3-Bromo-4-hydroxy-5-methoxycinnamic acid

Purity: 97% CAS No.: 6948-33-0 Molecular Formula: C10H9BrO4 Formula Weight: 273.09 Odour: Odourless Melting Point: 255-257° MDL No.: MFCD00017334

Formula: C₁₀H₉BrO₄
MW: 273.09 g/mol
Melting Pt: 255…257 °C
Storage Temperature: Ambient
MDL Number: MFCD00017334
CAS Number: 6948-33-0

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR