5,6-Dimethoxybenzimidazole ≥98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
5,6-Dimethoxy-1H-benzo[d]imidazole
H25942.06
H25942.03
ALFAH25942.06EA
0
SGD
ALFAH25942.06
ALFAH25942.03
5,6-Dimethoxybenzimidazole ≥98%
5,6-Dimethoxybenzimidazole
Purity: 98%
CAS No.: 72721-02-9
Molecular Formula: C9H10N2O2
Formula Weight: 178.19
Melting Point: 183-189°
MDL No.: MFCD00612461
Formula:
C₉H₁₀N₂O₂ MW: 178,19 g/mol Melting Pt: 183…189 °C Storage Temperature: Ambient |
MDL Number:
MFCD00612461 CAS Number: 72721-02-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...