5-Amino-2-(trifluoromethyl)benzimidazole ≥97%

Supplier: Thermo Scientific
Danger

B22770.03 B22770.06
ALFAB22770.03EA 0 SGD
ALFAB22770.03 ALFAB22770.06
5-Amino-2-(trifluoromethyl)benzimidazole ≥97%
5-Amino-2-(trifluoromethyl)benzimidazole

Purity: 97+% CAS No.: 3671-66-7 Molecular Formula: C8H6F3N3 Formula Weight: 201.15 Odour: Odourless Melting Point: 186-190° UN No.: 2811 MDL No.: MFCD01632182

Formula: C₈H₆F₃N₃
MW: 201,15 g/mol
Melting Pt: 186…190 °C
Storage Temperature: Ambient
MDL Number: MFCD01632182
CAS Number: 3671-66-7
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR