1-Acetylisatin ≥97%

Supplier: Thermo Scientific

Synonyms: 1-Acetylindoline-2,3-dione

L17597.03 L17597.06 L17597.14
ALFAL17597.03EA 58.3 SGD
ALFAL17597.03 ALFAL17597.06 ALFAL17597.14
1-Acetylisatin ≥97%
1-Acetylisatin

Purity: 97% CAS No.: 574-17-4 Molecular Formula: C10H7NO3 Formula Weight: 189.17 Melting Point: 141-143° Beilstein Reference: 113959

Formula: C₁₀H₇NO₃
MW: 189,17 g/mol
Melting Pt: 141…143 °C
Storage Temperature: Ambient
MDL Number: MFCD00158542
CAS Number: 574-17-4
EINECS: 209-368-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR