4-Chloro-2-(methylthio)pyrimidine ≥98%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
4-Chloro-2-methylthiopyrimidine
B21481.09
B21481.18
B21481.30
ALFAB21481.09EA
0
SGD
ALFAB21481.09
ALFAB21481.18
ALFAB21481.30
4-Chloro-2-(methylthio)pyrimidine ≥98%
4-Chloro-2-(methylthio)pyrimidine
Purity: 98%
CAS No.: 49844-90-8
Molecular Formula: C5H5ClN2S
Formula Weight: 160.63
Storage Temperature: -30°C to -10°C
Flash Point:
Formula:
C₅H₅ClN₂S MW: 160,63 g/mol Boiling Pt: 118…120 °C (9 mmHg) Melting Pt: ∼–2 °C Density: 1,380 Flash Pt: >110 °C (230 °F) |
MDL Number:
MFCD00006083 CAS Number: 49844-90-8 EINECS: 256-500-6 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...