(3,4-Dimethoxyphenylthio)acetic acid ≥97%
Supplier: Thermo Scientific
|
Warning
|
B20569.03
B20569.06
ALFAB20569.03EA
0
SGD
ALFAB20569.03
ALFAB20569.06
(3,4-Dimethoxyphenylthio)acetic acid ≥97%
(3,4-Dimethoxyphenylthio)acetic acid
Purity: 97%
CAS No.: 95735-63-0
Molecular Formula: C10H12O4S
Formula Weight: 228.26
Melting Point: 101-103°
MDL No.: MFCD00052112
Formula:
C₁₀H₁₂O₄S MW: 228,26 g/mol Melting Pt: 101…103 °C Storage Temperature: Ambient |
MDL Number:
MFCD00052112 CAS Number: 95735-63-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...