3-(Boc-amino)-3-(3-methoxyphenyl)propionic acid 99%
Supplier: Thermo Scientific
Synonyms:
3-((tert-Butoxycarbonyl)amino)-3-(3-methoxyphenyl)propanoic acid
L19807.03
L19807.MD
ALFAL19807.03EA
0
SGD
ALFAL19807.03
ALFAL19807.MD
3-(Boc-amino)-3-(3-methoxyphenyl)propionic acid 99%
3-(Boc-amino)-3-(3-methoxyphenyl)propionic acid
Purity: 99%
CAS No.: 284493-53-4
Molecular Formula: C15H21NO5
Formula Weight: 295.34
Storage Temperature: 2°C to 8°C
Melting Point: 81-83°
MDL No.: MFCD02090693
Formula:
C₁₅H₂₁NO₄ MW: 295,34 g/mol Melting Pt: 81…83 °C |
MDL Number:
MFCD02090693 CAS Number: 284493-53-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...