3-(Boc-amino)phenylboronic acid 95%

Supplier: Thermo Scientific
Warning

Synonyms: 3-Aminobenzeneboronic acid, N-BOC protected, 3-(N-Boc-amino)phenylboronic acid, (3-((tert-Butoxycarbonyl)amino)phenyl)boronic acid, 3-Boronoaniline, N-BOC protected, 3-[(tert-Butoxycarbonyl)amino]benzeneboronic acid, 3-(Boc-amino)benzeneboronic acid

H27258.06 H27258.03
ALFAH27258.06EA 0 SGD
ALFAH27258.06 ALFAH27258.03
3-(Boc-amino)phenylboronic acid 95%
3-(Boc-amino)phenylboronic acid

Purity: 95% CAS No.: 380430-68-2 Molecular Formula: C11H16BNO4 Formula Weight: 237.06 Melting Point: ca 155° dec. MDL No.: MFCD03411945

Formula: C₁₁H₁₆BNO₄
MW: 237,06 g/mol
Melting Pt: ∼ 155 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD03411945
CAS Number: 380430-68-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR