3-Chloro-4-[(trifluoromethyl)thio]aniline ≥97%

Supplier: Thermo Scientific
Danger

B23500.03 B23500.06
ALFAB23500.03EA 0 SGD
ALFAB23500.03 ALFAB23500.06
3-Chloro-4-[(trifluoromethyl)thio]aniline ≥97%
3-Chloro-4-[(trifluoromethyl)thio]aniline

Purity: 97% CAS No.: 64628-74-6 Molecular Formula: C7H5ClF3NS Formula Weight: 227.64 Melting Point: 41-44° Beilstein Reference: 6314200 UN No.: 2811 MDL No.: MFCD00190131

Formula: C₇H₅ClF₃NS
MW: 227,64 g/mol
Melting Pt: 41…44 °C
Storage Temperature: Ambient
MDL Number: MFCD00190131
CAS Number: 64628-74-6
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR