6-(Chloromethyl)uracil ≥98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
6-(Chloromethyl)pyrimidine-2,4(1H,3H)-dione
B21985.03
B21985.06
ALFAB21985.03EA
0
SGD
ALFAB21985.03
ALFAB21985.06
6-(Chloromethyl)uracil ≥98%
6-(Chloromethyl)uracil
Purity: 98%
CAS No.: 18592-13-7
Molecular Formula: C5H5ClN2O2
Formula Weight: 160.56
Melting Point: 256-258° dec.
MDL No.: MFCD00010095
Formula:
C₅H₅ClN₂O₂ MW: 160,56 g/mol Melting Pt: 256…258 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00010095 CAS Number: 18592-13-7 EINECS: 242-431-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...