5-Acetyluracil ≥98%
Supplier: Thermo Scientific
Synonyms:
5-Acetylpyrimidine-2,4(1H,3H)-dione
44378.03
44378.06
ALFA44378.03EA
0
SGD
ALFA44378.03
ALFA44378.06
5-Acetyluracil ≥98%
5-Acetyluracil
Purity: 98%
CAS No.: 6214-65-9
Molecular Formula: C6H6N2O3
Formula Weight: 154.11
Physical Form: Powder
Formula:
C₆H₆N₂O₃ MW: 154,11 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00151968 CAS Number: 6214-65-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...