4-Amino-4'-nitrostilbene-2,2'-disulphonic acid ≥80%, tech.
Supplier: Thermo Scientific
B21467.03
B21467.06
ALFAB21467.03EA
0
SGD
ALFAB21467.03
ALFAB21467.06
4-Amino-4'-nitrostilbene-2,2'-disulphonic acid ≥80%, tech.
4-Amino-4'-nitrostilbene-2,2'-disulphonic acid
Purity: 80%
CAS No.: 119-72-2
Molecular Formula: C14H12N2O8S2
Formula Weight: 400.38
Odour: Odourless
Melting Point:
Formula:
C₁₄H₁₂N₂O₈S₂ MW: 400,38 g/mol Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00035915 CAS Number: 119-72-2 EINECS: 204-346-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...