3-(Boc-amino)-4-methylphenylboronic acid ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: 3-(Boc-amino)-4-methylbenzeneboronic acid, 3-BOC-amino-4-methylphenylboronic acid, (3-((tert-Butoxycarbonyl)amino)-4-methylphenyl)boronic acid

H52548.MD H52548.03
ALFAH52548.MDEA 0 SGD
ALFAH52548.MD ALFAH52548.03
3-(Boc-amino)-4-methylphenylboronic acid ≥98%
3-(Boc-amino)-4-methylphenylboronic acid

Purity: 98% CAS No.: 850568-81-9 Molecular Formula: C12H18BNO4 Formula Weight: 251.09 Melting Point: 300° MDL No.: MFCD04115654

Formula: C₁₂H₁₈BNO₄
MW: 251,09 g/mol
Melting Pt: 300 °C
Storage Temperature: Ambient
MDL Number: MFCD04115654
CAS Number: 850568-81-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR