1,3-Bis(2-hydroxyhexafluoroisopropyl)benzene 96%

Supplier: Thermo Scientific
Warning

L16894.14 L16894.06
ALFAL16894.14EA 461 SGD
ALFAL16894.14 ALFAL16894.06
1,3-Bis(2-hydroxyhexafluoroisopropyl)benzene 96%
1,3-Bis(2-hydroxyhexafluoroisopropyl)benzene

Purity: 96% CAS No.: 802-93-7 Molecular Formula: C12H6F12O2 Formula Weight: 410.16 Flash Point: 97°(206°F) Refractive Index: 1.3890 Melting Point: 9-10° Boiling Point: 99-100°/20mm Beilstein Reference: 2024879 MDL No.: MFCD00042090

Formula: C₁₂H₆F₁₂O₂
MW: 410,16 g/mol
Boiling Pt: 99…100 °C (20 mmHg)
Melting Pt: 9…10 °C
Density: 1,659
Flash Pt: 97 °C (206 °F)
Storage Temperature: Ambient
MDL Number: MFCD00042090
CAS Number: 802-93-7
EINECS: 212-354-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR