Potassium(iodomethyl)trifluoroborate ≥90%, tech.
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Potassium trifluoro(iodomethyl)borate
H63839.03
H63839.06
ALFAH63839.03EA
138
SGD
ALFAH63839.03
ALFAH63839.06
Potassium(iodomethyl)trifluoroborate ≥90%, tech.
Potassium (iodomethyl)trifluoroborate
Formula:
CH₂BF₃IK MW: 247,83 g/mol Melting Pt: 242…258 °C Storage Temperature: Ambient |
MDL Number:
MFCD11112187 CAS Number: 888711-47-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...