2-Chloro-3-cyano-6-picoline ≥99%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
2-Chloro-3-cyano-6-methylpyridine, 2-Chloro-6-methylnicotinonitrile
A15648.03
A15648.06
A15648.14
ALFAA15648.03EA
0
SGD
ALFAA15648.03
ALFAA15648.06
ALFAA15648.14
2-Chloro-3-cyano-6-picoline ≥99%
2-Chloro-3-cyano-6-picoline
Purity: 99%
CAS No.: 28900-10-9
Molecular Formula: C7H5ClN2
Formula Weight: 152.58
Melting Point: 114-117°
Beilstein Reference: 120414
Formula:
C₇H₅ClN₂ MW: 152,58 g/mol Melting Pt: 114…117 °C Storage Temperature: Ambient |
MDL Number:
MFCD00082767 CAS Number: 28900-10-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...