3-Nitro-4-picoline ≥98%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
4-Methyl-3-nitropyridine
L07398.03
L07398.06
ALFAL07398.03EA
0
SGD
ALFAL07398.03
ALFAL07398.06
3-Nitro-4-picoline ≥98%
3-Nitro-4-picoline
Purity: 98%
CAS No.: 5832-44-0
Molecular Formula: C6H6N2O2
Formula Weight: 138.13
Storage Temperature: 2°C to 8°C
Flash Point: 108°(226°F)
Refractive Index: 1.5460
Melting Point: 18-22°
Boiling Point: 238°
Beilstein Reference: 118796
UN No.: 2810
MDL No.: MFCD00051829
Formula:
C₆H₆N₂O₂ MW: 138,13 g/mol Boiling Pt: 238 °C Melting Pt: 18…22 °C Density: 1,23 Flash Pt: 108 °C (226 °F) |
MDL Number:
MFCD00051829 CAS Number: 5832-44-0 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...