3-Nitro-4-(1-piperidinyl)benzaldehyde 96%
Supplier: Thermo Scientific
|
Warning
|
B22381.03
B22381.06
ALFAB22381.03EA
0
SGD
ALFAB22381.03
ALFAB22381.06
3-Nitro-4-(1-piperidinyl)benzaldehyde 96%
3-Nitro-4-(1-piperidinyl)benzaldehyde
Purity: 96%
CAS No.: 39911-29-0
Molecular Formula: C12H14N2O3
Formula Weight: 234.26
Melting Point: 50-54°
MDL No.: MFCD00831780
Formula:
C₁₂H₁₄N₂O₃ MW: 234,26 g/mol Melting Pt: 50…54 °C Storage Temperature: Ambient |
MDL Number:
MFCD00831780 CAS Number: 39911-29-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...