Methyltributylammonium bromide ≥98%
Supplier: Thermo Scientific
|
Warning
|
B23828.09
B23828.18
B23828.30
ALFAB23828.09EA
0
SGD
ALFAB23828.09
ALFAB23828.18
ALFAB23828.30
Methyltributylammonium bromide ≥98%
Methyltributylammonium bromide
Purity: 98%
CAS No.: 37026-88-3
Molecular Formula: [CH3(CH2)3]3NCH3Br
Formula Weight: 280.29
Melting Point: 120-122°
Beilstein Reference: 3704268
Formula:
C₉H₂₂BrN MW: 280,29 g/mol Melting Pt: 120…122 °C Storage Temperature: Ambient |
MDL Number:
MFCD00043201 CAS Number: 37026-88-3 EINECS: 253-313-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...