(R)-(-)-2-Methyl-2,4-pentanediol ≥98%

Supplier: Thermo Scientific
Warning

L14523.03 L14523.MD
ALFAL14523.03EA 265 SGD
ALFAL14523.03 ALFAL14523.MD
(R)-(-)-2-Methyl-2,4-pentanediol ≥98%
(R)-(-)-2-Methyl-2,4-pentanediol

Purity: 98+% CAS No.: 99210-90-9 Molecular Formula: C6H14O2 Formula Weight: 118.18 Flash Point: 93°(199°F) Refractive Index: 1.4275 Boiling Point: 197° Beilstein Reference: 6051650 Optical Rotation: -21° (neat) MDL No.: MFCD00064275

Formula: C₆H₁₄O₂
MW: 118,18 g/mol
Boiling Pt: 197 °C
Density: 0,920
Flash Pt: 93 °C (199 °F)
Storage Temperature: Ambient
MDL Number: MFCD00064275
CAS Number: 99210-90-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR