1-Tritylimidazole ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: Clotrimazole Impurity F, 1-Trityl-1H-imidazole

L17694.03 L17694.06
ALFAL17694.03EA 0 SGD
ALFAL17694.03 ALFAL17694.06
1-Tritylimidazole ≥98%
1-Tritylimidazole

Purity: 98% CAS No.: 15469-97-3 Molecular Formula: C22H18N2 Formula Weight: 310.40 Odour: Odourless Melting Point: 220-224° Beilstein Reference: 238171 MDL No.: MFCD00229427

Formula: C₂₂H₁₈N₂
MW: 310,40 g/mol
Melting Pt: 220…224 °C
Storage Temperature: Ambient
MDL Number: MFCD00229427
CAS Number: 15469-97-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR