5-Fluoronicotinonitrile ≥97%

Supplier: Thermo Scientific
Danger

Synonyms: 3-Cyano-5-fluoropyridine

H34292.MD H34292.03
ALFAH34292.MDEA 0 SGD
ALFAH34292.MD ALFAH34292.03
5-Fluoronicotinonitrile ≥97%
5-Fluoronicotinonitrile

Purity: 97% CAS No.: 696-42-4 Molecular Formula: C6H3FN2 Formula Weight: 122.10 Storage Temperature: 2°C to 8°C Melting Point: 50-54° MDL No.: MFCD09027295

Formula: C₆H₃FN₂
MW: 122,10 g/mol
Melting Pt: 50…54 °C
MDL Number: MFCD09027295
CAS Number: 696-42-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR