2,4-Dimethyl-3-heptanone ≥99%

Supplier: Thermo Scientific
Warning

B20384.09 B20384.18
ALFAB20384.09EA 0 SGD
ALFAB20384.09 ALFAB20384.18
2,4-Dimethyl-3-heptanone ≥99%
2,4-Dimethyl-3-heptanone

Purity: 99% CAS No.: 18641-71-9 Molecular Formula: C9H18O Formula Weight: 142.24 Flash Point: 44°(111°F) Odour: Pleasant Boiling Point: 170° UN No.: 1224 MDL No.: MFCD00027146

Formula: C₉H₁₈O
MW: 142,24 g/mol
Boiling Pt: 170 °C
Density: 0,812
Flash Pt: 44 °C (111 °F)
Storage Temperature: Ambient
MDL Number: MFCD00027146
CAS Number: 18641-71-9
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR