Perfluoro(allylbenzene) 97%
Supplier: Thermo Scientific
B21469.06
B21469.14
ALFAB21469.06EA
0
SGD
ALFAB21469.06
ALFAB21469.14
Perfluoro(allylbenzene) 97%
Perfluoro(allylbenzene)
Perfluoro(allylbenzene) is used as a pharmaceutical intermediate and as a chemical in organic synthesis.
Formula:
C₉F₁₀ MW: 298,08 g/mol Density: 1,708 Storage Temperature: Ambient |
MDL Number:
MFCD00153225 CAS Number: 67899-41-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...