1,3-Bis(methylthio)propane 97

Supplier: Thermo Fisher Scientific

B22397.03 B22397.06
ALFAB22397.03EA 0 SGD
ALFAB22397.03 ALFAB22397.06
1,3-Bis(methylthio)propane 97
1,3-Bis(methylthio)propane

Purity: 97% CAS No.: 24949-35-7 Molecular Formula: CH3S(CH2)3SCH3 Formula Weight: 136.28 Flash Point: 100°(212°F) Refractive Index: 1.5190 Odour: Stench Boiling Point: 88-90°/12mm Beilstein Reference: 1732333 MDL No.: MFCD00039811

Formula: C₅H₁₂S₂
MW: 136.28 g/mol
Boiling Pt: 88…90 °C (12 mmHg)
Density: 1.011
Flash Pt: >100 °C (212 °F)
Storage Temperature: Ambient
MDL Number: MFCD00039811
CAS Number: 24949-35-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR