(R)-3-(Boc-amino)-3-phenylpropionic acid 95%

Supplier: Thermo Scientific

Synonyms: (R)-3-((tert-Butoxycarbonyl)amino)-3-phenylpropanoic acid, (R)-N-BOC-3-Amino-3-phenylpropanoic acid

H51984.MD H51984.03
ALFAH51984.MDEA 303 SGD
ALFAH51984.MD ALFAH51984.03
(R)-3-(Boc-amino)-3-phenylpropionic acid 95%
(R)-3-(Boc-amino)-3-phenylpropionic acid

Purity: 95% CAS No.: 161024-80-2 Molecular Formula: C14H19NO4 Formula Weight: 265.31 Beilstein Reference: 5404389 MDL No.: MFCD01320859

Formula: C₁₄H₁₉NO₄
MW: 265,31 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01320859
CAS Number: 161024-80-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR