Lead titanate (30% Oxide) polymeric precursor

Supplier: Thermo Scientific
Danger

Synonyms: Lead titanium oxide, Lead(II) titanium oxide, Lead(II) titanate, Lead titanate (IV), Lead titanate

39756.09 39756.04
ALFA39756.09EA 0 SGD
ALFA39756.09 ALFA39756.04
Lead titanate (30% Oxide) polymeric precursor
Lead titanate

CAS No.: 12060-00-3 Flash Point: 12°(54°F) Physical Form: Viscous liquid Sensitivity: Moisture Sensitive Appearance: Brown Solubility: Soluble in alcohols MDL No.: MFCD00011159


Used in electronic devices

Formula: O₃PbTi
MW: 303,08 g/mol
Melting Pt: 1120 °C
Density: 7,5 g/cm³ (20 °C)
Flash Pt: 12 °C (54 °F)
Storage Temperature: Ambient
MDL Number: MFCD00011159
CAS Number: 12060-00-3
EINECS: 235-038-9
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR