2-Methyl-2-propen-1-ol ≥98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
3-Hydroxy-2-methylpropene, 2-Methyl-2-propen-1-ol, 2-Methyl allyl alcohol, 2-Methylprop-2-en-1-ol
H60502.03
H60502.06
ALFAH60502.03EA
28.6
SGD
ALFAH60502.03
ALFAH60502.06
2-Methyl-2-propen-1-ol ≥98%
2-Methyl-2-propen-1-ol
Formula:
C₄H₈O MW: 72,11 g/mol Boiling Pt: 113…115 °C Density: 0,857 Flash Pt: 32 °C (90 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004737 CAS Number: 513-42-8 EINECS: 208-161-0 UN: 2614 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...