Centrifuge bottles, PP/PC, with screw cap, Corning®
Supplier: Corning
431840 431841 431843 431844 431845 431846 431847
CORN431840EA 1130 SGD
CORN431840 CORN431841 CORN431843 CORN431844 CORN431845 CORN431846 CORN431847
Centrifuge bottles, PP/PC, with screw cap, Corning®
Bottles Centrifuge Bottles
PP, translucent or PC, clear.
  • Clear graduations and textured marking spot
  • Specially designed cap with a built-in easy grip feature
  • Caps are not interchangeable among different size/material bottles
  • Extra-thick walls for durability
  • Wide-mouth bottle design for improved liquid handling
Centrifuge bottles for large volume processing in cell harvesting, pelleting and protein purification. Allow researchers to decrease the number of centrifugation cycles, improve the laboratory efficiency and boost productivity. Two cap styles: standard and high performance with silicone O-ring. Leak-free processing at maximum rated speeds (Fill volume should not exceed 80% of the total bottle capacity).