Semi-dry blotters
Supplier: CLEAVER SCIENTIFIC
SD10 SD20
CLEVSD10EA 984 SGD
CLEVSD10 CLEVSD20
Semi-dry blotters
Blotting Apparatus
These semi-dry blotters offer rapid transfer times for DNA, RNA and protein blotting – typically 15 to 30 minutes. All units can be used for all types of blotting: Western, Southern and Northern via uncomplicated buffer and set-up procedures and are compatible with gel thicknesses from 0,25 mm up to 10 mm without the need for additional equipment. Each unit is compatible with their respective omniPAGE vertical gel system. The electrodes, comprising platinum coated anode and stainless steel cathode exhibits no corrosion.
  • Suitable for Western, Southern, and Northern blots
  • Rapid transfer times
  • Uniform heat dispersion
  • Variable gel thickness
  • Minimal buffer volume