Filtering flasks, micro, with glass hose connection, PYREX®
Supplier: Corning
5360-125 5360-25 5360-50
CORN5360-125EA 28.9 SGD
CORN5360-125 CORN5360-25 CORN5360-50
Filtering flasks, micro, with glass hose connection, PYREX®
Flasks Filter Flasks
Made of PYREX® borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Flasks have a beaded top that provides an excellent fit for rubber stoppers. These small filtering flasks are recommended for microchemical use. All flasks have permanent white enamel marking spots.