Test tubes, glass, with rim, PYREX® VISTA™
Supplier: Corning
70800-10 70800-12 70800-13 70800-15 70800-16 70800-18 70800-20 70800-25 70800-25X
CORN70800-10EA 85.4 SGD
CORN70800-10 CORN70800-12 CORN70800-13 CORN70800-15 CORN70800-16 CORN70800-18 CORN70800-20 CORN70800-25 CORN70800-25X
Test tubes, glass, with rim, PYREX® VISTA™
Tubes Test Tubes
Annealed test tubes offer excellent heat resistance and chemical stability.
  • Non-sterile
Reusable tubes have a round-bottom design and feature fire-polished rims. All tubes except the 55 and 70 ml sizes have a blue enamel marking spot.
Delivery information: Supplied without caps