Powder funnels, glass, PYREX®
Supplier: Corning
6220-65 6220-100 6220-125 6220-150 6220-75
CORN6220-65EA 392 SGD
CORN6220-65 CORN6220-100 CORN6220-125 CORN6220-150 CORN6220-75
Powder funnels, glass, PYREX®
Funnels Powder Funnels
PYREX®, borosilicate glass.
With short, wide stem. Used for transferring powders or filling bottles.
Order Now
SPECIFICATIONS
Environmentally Preferable