Stopcock, straight bore, PYREX®
Supplier: Corning
7280-2 7280-4
CORN7280-2EA 422 SGD
CORN7280-2 CORN7280-4
Stopcock, straight bore, PYREX®
Stopcocks
PYREX®, borosilicate glass with glass key.
Straight bore, side arm seals are heavy and it can be used at pressure down to 10 torr.