Culture tubes with screw caps, PYREX® VISTA™
Supplier: Corning
70825-13 70825-16 70825-16X 70825-16XX 70825-20X 70825-20 70825-25
CORN70825-13EA 215 SGD
CORN70825-13 CORN70825-16 CORN70825-16X CORN70825-16XX CORN70825-20X CORN70825-20 CORN70825-25
Culture tubes with screw caps, PYREX® VISTA™
Tubes Culture Tubes
Borosilicate glass, reusable.
Tubes have large blue enamel marking spots for simple identification.
Delivery information: Supplied with deep form phenolic screw caps with a rubber liner to facilitate handling and sealing after autoclaving.
Order Now
SPECIFICATIONS
Environmentally Preferable