Stoppers for reagent bottle and ground joints, PYREX®
Supplier: Corning
7575-14 7575-24 7575-29
CORN7575-14EA 88.3 SGD
CORN7575-14 CORN7575-24 CORN7575-29
Stoppers for reagent bottle and ground joints, PYREX®
Stoppers Glass Stoppers
Combination glass stoppers from PYREX®, borsilicate glass with closed bottoms may be used as [ST] joint stoppers and as reagent bottle stoppers. Barrel-shaped head is easy to clean and rotate, side grooves prevent slippage, and flat top allows stopper to stand on head.
  • Lightweight, but strong enough to keep chipping and breakage to a minimum
Order Now
SPECIFICATIONS
Environmentally Preferable