Flasks, flat bottom, long neck/short neck, PYREX®
Supplier: Corning
4060-125 4060-12L 4060-1L 4060-250 4060-2L 4060-50 4060-500 4060-6L
CORN4060-125EA 67.9 SGD
CORN4060-125 CORN4060-12L CORN4060-1L CORN4060-250 CORN4060-2L CORN4060-50 CORN4060-500 CORN4060-6L
Flasks, flat bottom, long neck/short neck, PYREX®
Flasks Flat Bottom Flasks
Manufactured from borosilicate glass.
  • High thermal shock resistance
  • Provide contamination free results
  • High chemical durability
Tooled mouth ensures uniform stopper fit. Florence, long neck. Normal working temperature is 230 °C (446 °F).